Natriumacetat-[18O2] (95% ¹⁸O), Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ200
SIAL487805-1GEA
1670
EUR
SIAL487805-1G
Natriumacetat-[18O2] (95% ¹⁸O), Sigma-Aldrich®
Natriumacetat-[18O2]
Formel:
H₃CCOONa Lagertemperatur: Raumtemperatur |
MDL:
MFCD00145293 CAS-Nummer: 66012-98-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...