(Trimethylsilyl)diazomethan 2 mol/l in Diethylether, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
SIAL527254-100MLEA
461
EUR
SIAL527254-100ML
(Trimethylsilyl)diazomethan 2 mol/l in Diethylether, Sigma-Aldrich®
(Trimethylsilyl)diazomethan
Formel:
C₄H₁₀N₂Si Molecular Weight: 114,22 g/mol |
MDL:
MFCD00053946 UN: 1992 ADR: 3,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...