Carbamazepin
Lieferant: LGC Standards PROMOCHEM
PROOMM0076.00EA
229
EUR
PROOMM0076.00
Carbamazepin
Carbamazepin
Formel:
C₁₅H₁₂N₂O Molecular Weight: 236,27 |
MDL:
MFCD00005073 CAS-Nummer: 298-46-4 EINECS: 206-062-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...