Carbamazepin

Lieferant: Molekula

MOLEM18994181EA 0 EUR
MOLEM18994181
Carbamazepin
Carbamazepin
Formel: C₁₅H₁₂N₂O
Molecular Weight: 236,27 g/mol
MDL: MFCD00005073
CAS-Nummer: 298-46-4
EINECS: 206-062-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR