2-Methyl-4'-(methylthio)-2-morpholinopropiophenon, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL405639-50GEA
132
EUR
SIAL405639-50G
2-Methyl-4'-(methylthio)-2-morpholinopropiophenon, Sigma-Aldrich®
2-Methyl-4'-(methylthio)-2-morpholinopropiophenon
Formel:
C₁₅H₂₁NO₂S |
MDL:
MFCD00083014 CAS-Nummer: 71868-10-5 EINECS: 400-600-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...