3-(4-Morpholinyl)propiophenon Hydrochlorid ≥98%

Lieferant: Thermo Scientific
Achtung

B23686.22EA 186 EUR
B23686.22 B23686.06 B23686.14
3-(4-Morpholinyl)propiophenon Hydrochlorid ≥98%
3-(4-Morpholinyl)propiophenon Hydrochlorid
Formel: C₁₃H₁₈ClNO₂
Schmelzpunkt: 180…182 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00035332
CAS-Nummer: 1020-16-2
EINECS: 213-815-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR