p-Terphenyl-[D14]
Lieferant: EHRENSTORFER GMBH
EHERC20935300EA
47
EUR
EHERC20935300
p-Terphenyl-[D14]
p-Terphenyl-[D14]
Lagertemperatur:
Raumtemperatur |
CAS-Nummer:
1718-51-0 UN: 3077 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...