p-Terphenyl-[D14] (98% D)

Lieferant: LGC Standards PROMOCHEM

PROOCIL-DLM-382-5EA 1497 EUR
PROOCIL-DLM-382-5 PROOCIL-DLM-382-1
p-Terphenyl-[D14] (98% D)
p-Terphenyl-[D14]
Lagertemperatur: Raumtemperatur
CAS-Nummer: 1718-51-0
UN: 3077
ADR: 9,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR