4-Fluorphenylchlorformiat 98%
|
Gefahr
|
ACRO308320050EA
108
EUR
ACRO308320050
ACRO308320010
4-Fluorphenylchlorformiat 98%
4-Fluorphenylchlorformiat
Formel:
C₇H₄ClFO₂ Molecular Weight: 174,5569 g/mol Flammpunkt: 76 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00013257 CAS-Nummer: 38377-38-7 UN: 3277 ADR: 6.1,II |
Spezifikation Testergebnisse
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4923 to 1.4943 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...