N-(2-Hydroxyethyl)iminodiessigsäure, Sigma-Aldrich®

Lieferant: Merck

SIAL54345-5GEA 41 EUR
SIAL54345-5G
N-(2-Hydroxyethyl)iminodiessigsäure, Sigma-Aldrich®
N-(2-Hydroxyethyl)iminodiessigsäure
Formel: C₆H₁₁NO₅
MDL: MFCD00004293
CAS-Nummer: 93-62-9
EINECS: 202-263-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR