N-Bromacetamid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ200

SIALB2377-50GEA 443 EUR
SIALB2377-50G
N-Bromacetamid, Sigma-Aldrich®
N-Bromacetamid
Formel: C₂H₄BrNO
Molecular Weight: 137,97 g/mol
MDL: MFCD00037097
CAS-Nummer: 79-15-2
EINECS: 201-181-0
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR