2',4'-Dimethoxy-2,2,2-trifluoracetophenon, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL514659-1GEA 198 EUR
SIAL514659-1G
2',4'-Dimethoxy-2,2,2-trifluoracetophenon, Sigma-Aldrich®
2',4'-Dimethoxy-2,2,2-trifluoracetophenon
Formel: C₁₀H₉F₃O₃
Molecular Weight: 234,1719 g/mol
MDL: MFCD00835330
CAS-Nummer: 578-16-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR