5'-Brom-2'-hydroxy-3'-nitropropiophenon

Lieferant: CHEMPUR

CHMPFL12098.POREA 0 EUR
CHMPFL12098.POR
5'-Brom-2'-hydroxy-3'-nitropropiophenon
5'-Brom-2'-hydroxy-3'-nitropropiophenon
Formel: C₉H₈BrNO₄
Molecular Weight: 274,0681 g/mol
MDL: MFCD01067741
CAS-Nummer: 90725-67-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR