4-Formyl-3-methoxyphenylboronsäure
Lieferant: COMBI-BLOCKS INC.
COBBFA-1835-1GEA
478
EUR
COBBFA-1835-1G
COBBFA-1835-5G
COBBFA-1835-250M
4-Formyl-3-methoxyphenylboronsäure
4-Formyl-3-methoxyphenylboronsäure
Formel:
C₈H₉BO₄ |
MDL:
MFCD10697427 CAS-Nummer: 815620-00-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...