Cyclopentadienylzirkoniumtrichlorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL475203-1GEA 68.2 EUR
SIAL475203-1G
Cyclopentadienylzirkoniumtrichlorid, Sigma-Aldrich®
Cyclopentadienylzirkoniumtrichlorid
Formel: C₅H₅Cl₃Zr
Molecular Weight: 262,676 g/mol
MDL: MFCD00015756
CAS-Nummer: 34767-44-7
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR