Pentafluorphenyl-5-(dimethylamino)-1-naphthalinsulfonat

Lieferant: BIONET RESEARCH (CHEMPUR)

BIOT1Y-0265.500EA 0 EUR
BIOT1Y-0265.500 BIOT1Y-0265.1 BIOT1Y-0265.10 BIOT1Y-0265.5
Pentafluorphenyl-5-(dimethylamino)-1-naphthalinsulfonat
Pentafluorphenyl-5-(dimethylamino)-1-naphthalinsulfonat
Formel: C₁₈H₁₂F₅NO₃S
MDL: MFCD06659540
CAS-Nummer: 663175-92-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR