Methyl-4-ethinylbenzoat, Sigma-Aldrich®

Lieferant: Merck

SIAL724653-1GEA 103 EUR
SIAL724653-1G
Methyl-4-ethinylbenzoat, Sigma-Aldrich®
Methyl-4-ethinylbenzoat
Formel: C₁₀H₈O₂
Molecular Weight: 160,17 g/mol
MDL: MFCD00168820
CAS-Nummer: 3034-86-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR