Tubing connectors, reducing, TPX
Lieferant: Semadeni
229-4641EA 30.8 EUR
229-4641 229-4642
Tubing connectors, reducing, TPX
Fittings und Verbinder
Reducing connectors, made of TPX, transparent. Strong design with long, conical connections.
  • Temperature resistant up to 130 °C
  • Sterilisable
  • Sturdy