Precision column holder
Lieferant: Cytiva
17-1455-01EA 818.73 EUR
17-1455-01
Precision column holder
Vorsäulenhalter
Adapts PC columns designed for SMART System and for use with other high performance chromatography systems classic ÄKTA systems.
  • Low dead volume fittings maintain resolution
Lieferumfang: Includes column holders for 30 to 50 mm and 300 mm long PC columns, end fittings, tubing connectors/M6, screw caps and stop plugs.