TES-Natriumsalz (2-(Tris(hydroxymethyl)methylamino)ethansulfonsäure Natriumsalz)
Lieferant: Thermo Scientific
J62706.14EA
49.09
EUR
J62706.14
TES-Natriumsalz (2-(Tris(hydroxymethyl)methylamino)ethansulfonsäure Natriumsalz)
TES-Natriumsalz (2-(Tris(hydroxymethyl)methylamino)ethansulfonsäure Natriumsalz)
A zwitterionic Good's Buffer
Formel:
C₆H₁₄NNaO₆S Molecular Weight: 251,24 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00065482 CAS-Nummer: 70331-82-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...