2-Fluor-m-xylol

Lieferant: COMBI-BLOCKS INC.

Synonyme: 1-Fluor-2,6-dimethylbenzol2-Fluor-1,3-dimethylbenzol

COBBSS-4323-100GEA 374 EUR
COBBSS-4323-100G
2-Fluor-m-xylol
2-Fluor-m-xylol
Formel: C₈H₉F
Molecular Weight: 124,16 g/mol
Siedepunkt: 165…167 °C (1013 hPa)
Dichte: 0,988 g/cm³ (20 °C)
MDL: MFCD00039215
CAS-Nummer: 443-88-9
EINECS: 207-144-5
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR