4-Fluor-7-nitrobenzofurazan

Lieferant: Thermo Scientific
Achtung

J61336.MCEA 471 EUR
J61336.MC
4-Fluor-7-nitrobenzofurazan
4-Fluor-7-nitrobenzofurazan

Useful for amino acid and amine fluorescent labeling in HPLC

Formel: C₆H₂FN₃O₃
Molecular Weight: 183,1 g/mol
Schmelzpunkt: 53…57 °C
Flammpunkt: 110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00010196
CAS-Nummer: 29270-56-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR