Calciumcarbonat, Pulver, BAKER ANALYZED® ACS, J.T.Baker®

Synonyme: Marmor

1294-01EA 192.5 EUR
1294-01
Calciumcarbonat, Pulver, BAKER ANALYZED® ACS, J.T.Baker®
Calciumcarbonat
Formel: CaCO₃
Molecular Weight: 100,09 g/mol
Schmelzpunkt: 825 °C
Dichte: 2,71 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00010906
CAS-Nummer: 471-34-1
EINECS: 207-439-9
Merck Index: 13,01658

Bestellen

Spezifikation Testergebnisse

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (CaCO₃) (dried basis) ≥99.0 %
Insoluble in Dilute HCl ≤0.01 %
Ammonium (NH₄) ≤0.003 %
Chloride (Cl) ≤0.001 %
Fluoride (F) ≤0.0015 %
Sulfate (SO₄) ≤0.005 %
Barium (Ba) ≤0.01 %
Heavy Metals (as Pb) ≤0.001 %
Iron (Fe) ≤0.002 %
Magnesium (Mg) ≤0.01 %
Potassium (K) ≤0.01 %
Sodium (Na) ≤0.010 %
Strontium (Sr) ≤0.1 %
SGRM Lot2 - Average Particle Diameter, µm (APD), For Information Only
Specific Surface Area, m²/g, For Information Only
Bulk Density (g/cc), For Information Only
Mesh (Wet Screen Analysis) - On U.S. No. 325 Sieve, For Information Only

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR