2-Brom-1,1,1-trifluorethan

Lieferant: SIA ENAMINE

ENAMEN300-77137.1GEA 0 EUR
ENAMEN300-77137.1G
2-Brom-1,1,1-trifluorethan
2-Brom-1,1,1-trifluorethan
Formel: F₃CCH₂Br
CAS-Nummer: 421-06-7
EINECS: 207-001-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR