4-Fluor-3-methyl-5-(trifluormethyl)benzylamin 97%

Lieferant: Alfa Aesar
Gefahr

H33499.MDEA 41.9 EUR
H33499.MD H33499.03
4-Fluor-3-methyl-5-(trifluormethyl)benzylamin 97%
4-Fluor-3-methyl-5-(trifluormethyl)benzylamin
Formel: C₉H₉F₄N
Lagertemperatur: Raumtemperatur
MDL: MFCD22201070
CAS-Nummer: 1373921-00-6
UN: 3267
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR