2,4-Dichlor-L-phenylalanin

Lieferant: COMBI-BLOCKS INC.

COBBSS-2911-1GEA 33.1 EUR
COBBSS-2911-1G
2,4-Dichlor-L-phenylalanin
2,4-Dichlor-L-phenylalanin
Formel: C₉H₉Cl₂NO₂
Molecular Weight: 234,1 g/mol
MDL: MFCD01860882
CAS-Nummer: 111119-36-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR