2,4-Dichlor-D-phenylalanin ≥98%

H27269.03EA 146 EUR
H27269.03 H27269.MD
2,4-Dichlor-D-phenylalanin ≥98%
2,4-Dichlor-D-phenylalanin
Formel: C₉H₉Cl₂NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01860873
CAS-Nummer: 114872-98-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR