3,5-Difluor-DL-phenylalanin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL546550-500MGEA 45.2 EUR
SIAL546550-500MG
3,5-Difluor-DL-phenylalanin, Sigma-Aldrich®
3,5-Difluor-DL-phenylalanin
Formel: C₉H₉F₂NO₂
MDL: MFCD00061308
CAS-Nummer: 32133-37-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR