Dibenzyldicarbonat, Sigma-Aldrich®

Lieferant: Merck

SIAL311219-25GEA 305 EUR
SIAL311219-25G SIAL311219-5G
Dibenzyldicarbonat, Sigma-Aldrich®
Dibenzyldicarbonat
Formel: C₁₆H₁₄O₅
MDL: MFCD00043124
CAS-Nummer: 31139-36-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR