4-Chlor-6-methoxy-2-methylchinolin

Lieferant: CHEMPUR

MAYBBTB10032.1EA 0 EUR
MAYBBTB10032.1 CHMPBTB10032.250
4-Chlor-6-methoxy-2-methylchinolin
4-Chloro-6-methoxy-2-methylquinoline
Formel: C₁₁H₁₀ClNO
MDL: MFCD00093863
CAS-Nummer: 50593-73-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR