2-Isopropylamino-3'-methoxyacetophenon

Lieferant: Thermo Scientific

H50736.MDEA 67.8 EUR
H50736.MD
2-Isopropylamino-3'-methoxyacetophenon
2-Isopropylamino-3'-methoxyacetophenon
Formel: C₁₂H₁₇NO₂
MDL: MFCD08064238
CAS-Nummer: 857724-35-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR