4-Chlor-2-(trifluormethoxy)benzylbromid ≥97%

Lieferant: Thermo Scientific

Gefahr

H33801.MDEA 45.1 EUR
H33801.MD H33801.03
4-Chlor-2-(trifluormethoxy)benzylbromid ≥97%
4-Chlor-2-(trifluormethoxy)benzylbromid
Formel: C₈H₅BrClF₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD18393833
CAS-Nummer: 1261606-06-7
UN: 3265
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR