Molybdänborid, Sigma-Aldrich®

Lieferant: Merck

709182-10G
SIAL709182-10GEA 108 EUR
SIAL709182-10G
Molybdänborid, Sigma-Aldrich®
Molybdenum boride
Formel: BMo
Molecular Weight: 106,75 g/mol
MDL: MFCD00014219
CAS-Nummer: 12006-98-3
EINECS: 234-492-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR