4'-(1-Piperazinyl)acetophenon, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL136468-5GEA 41.7 EUR
SIAL136468-5G
4'-(1-Piperazinyl)acetophenon, Sigma-Aldrich®
4'-(1-Piperazinyl)acetophenon
Formel: C₁₂H₁₆N₂O
Molecular Weight: 204,27
MDL: MFCD00005962
CAS-Nummer: 51639-48-6
EINECS: 257-332-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR