4'-(1-Piperazinyl)acetophenon 94%

Gefahr

ACRO159570050EA 31.2 EUR
ACRO159570050
4'-(1-Piperazinyl)acetophenon 94%
4'-(1-Piperazinyl)acetophenon
Formel: C₁₂H₁₆N₂O
Molecular Weight: 204,27
Schmelzpunkt: 107…110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005962
CAS-Nummer: 51639-48-6
EINECS: 257-332-6
UN: 3259
ADR: 8,III

Bestellen

Spezifikation Testergebnisse

Appearance Yellow Powder and/or chunks
Infrared spectrum Conforms
Titration with HClO4 ≥92.0 %
pH 4.1 to 4.5
Titration with HClO4 97.5 to 103 %
Heavy metals (as Pb) ≤0.002 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR