Mangan(II)sulfid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL400947-5GEA 80.6 EUR
SIAL400947-5G
Mangan(II)sulfid, Sigma-Aldrich®
Manganese(II) sulphide
Formel: MnS
Molecular Weight: 87,00 g/mol
MDL: MFCD00014210
CAS-Nummer: 18820-29-6
EINECS: 242-599-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR