trans-1,4-Cyclohexandicarbonsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIALC100757-5GEA 78.4 EUR
SIALC100757-5G
trans-1,4-Cyclohexandicarbonsäure, Sigma-Aldrich®
trans-1,4-Cyclohexanedicarboxylic acid
Formel: C₈H₁₂O₄
Molecular Weight: 172,18
MDL: MFCD00066203
CAS-Nummer: 619-82-9
EINECS: 210-614-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR