2,2,2,3'-Tetrafluoracetophenon, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL402702-10GEA 194 EUR
SIAL402702-10G
2,2,2,3'-Tetrafluoracetophenon, Sigma-Aldrich®
2,2,2,3'-Tetrafluoracetophenon
Formel: C₈H₄F₄O
Molecular Weight: 192,11 g/mol
MDL: MFCD00039236
CAS-Nummer: 708-64-5
EINECS: 211-903-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR