3-Fluoro-N-n-pentylbenzylamine, 97%

Lieferant: Thermo Scientific
H57919.03EA 306 EUR
ALFAH57919.03 ALFAH57919.MD
3-Fluoro-N-n-pentylbenzylamine, 97%
Purity: 97%
CAS No.: 90389-88-1
Molecular Formula: C12H18FN
Formula Weight: 195.28
MDL No.: MFCD11141097
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR