Phenylformiat, Sigma-Aldrich®

Lieferant: Merck

SIAL06556-25MLEA 359 EUR
SIAL06556-25ML SIAL06556-5ML
Phenylformiat, Sigma-Aldrich®
Phenylformiat
Formel: C₇H₆O₂
Molecular Weight: 122,12 g/mol
MDL: MFCD00014126
CAS-Nummer: 1864-94-4
EINECS: 217-471-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR