4',5'-Dimethoxy-2'-hydroxyacetophenon, Sigma-Aldrich®

Lieferant: Merck

SIALH9904-5GEA 208 EUR
SIALH9904-5G
4',5'-Dimethoxy-2'-hydroxyacetophenon, Sigma-Aldrich®
4',5'-Dimethoxy-2'-hydroxyacetophenon
Formel: C₁₀H₁₂O₄
MDL: MFCD00017241
CAS-Nummer: 20628-06-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR