3',5'-Dibenzyloxyacetophenon, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL215341-50GEA
186
EUR
SIAL215341-50G
3',5'-Dibenzyloxyacetophenon, Sigma-Aldrich®
3',5'-Dibenzyloxyacetophenon
Formel:
C₂₂H₂₀O₃ Molecular Weight: 332,40 g/mol |
MDL:
MFCD00004777 CAS-Nummer: 28924-21-2 EINECS: 249-315-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...