3',5'-Dichlor-2'-hydroxyacetophenon, Sigma-Aldrich®

Lieferant: Merck

SIAL383392-25GEA 285 EUR
SIAL383392-25G
3',5'-Dichlor-2'-hydroxyacetophenon, Sigma-Aldrich®
3',5'-Dichlor-2'-hydroxyacetophenon
Formel: C₈H₆Cl₂O₂
Molecular Weight: 205,04 g/mol
MDL: MFCD00016417
CAS-Nummer: 3321-92-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR