3',5'-Dichlor-2'-hydroxyacetophenon

Lieferant: CHEMPUR

CHMPFL12072.POREA 0 EUR
FLUO012072.5G CHMPFL12072.POR FLUO012072.25G
3',5'-Dichlor-2'-hydroxyacetophenon
3',5'-Dichlor-2'-hydroxyacetophenon
Formel: C₈H₆Cl₂O₂
Molecular Weight: 205,04 g/mol
MDL: MFCD00016417
CAS-Nummer: 3321-92-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR