D(+)-Galactosamin Hydrochlorid

Lieferant: U.S. Pharmacopeia

USPH1287722EA 432 EUR
USPH1287722
D(+)-Galactosamin Hydrochlorid
D(+)-Galactosamin Hydrochlorid
Formel: C₆H₁₄ClNO₅
Molecular Weight: 215,64
MDL: MFCD00135830
CAS-Nummer: 1772-03-8
EINECS: 217-198-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR