5'-Fluor-2'-methoxyacetophenon, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL683299-1GEA 74 EUR
SIAL683299-1G
5'-Fluor-2'-methoxyacetophenon, Sigma-Aldrich®
5'-Fluor-2'-methoxyacetophenon
Formel: C₉H₉FO₂
Molecular Weight: 168,17 g/mol
MDL: MFCD00671763
CAS-Nummer: 445-82-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR