2-Chloro-6-(trifluoromethyl)benzylamine 97%
|
Gefahr
|
H32981.03EA
198
EUR
H32981.03
H32981.MD
2-Chloro-6-(trifluoromethyl)benzylamine 97%
2-Chloro-6-(trifluoromethyl)benzylamine
Formel:
C₈H₇ClF₃N Molecular Weight: 209,596 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06213040 CAS-Nummer: 771582-27-5 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...