4'-Acetamidoacetophenon, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL631361-5GEA
44.7
EUR
SIAL631361-5G
SIAL631361-25G
4'-Acetamidoacetophenon, Sigma-Aldrich®
4'-Acetamidoacetophenon
Formel:
C₁₀H₁₁NO₂ Molecular Weight: 177,20 |
MDL:
MFCD00014965 CAS-Nummer: 2719-21-3 EINECS: 220-320-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...