Nickelsulfid, Sigma-Aldrich®

Lieferant: Merck
Gefahr


M-Clarity™ quality level = MQ100

SIAL343226-25GEA 189 EUR
SIAL343226-25G
Nickelsulfid, Sigma-Aldrich®
Nickelsulfid
Formel: Ni₃S₂
Molecular Weight: 240,21 g/mol
MDL: MFCD00217044
CAS-Nummer: 12035-72-2
EINECS: 234-829-6
UN: 3077
ADR: 9,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR