5-Fluor-2-methoxybenzylamin 97%, JRD

Gefahr

H32897.MDEA 59.4 EUR
H32897.MD H32897.03
5-Fluor-2-methoxybenzylamin 97%, JRD
5-Fluor-2-methoxybenzylamin
Formel: C₈H₁₀FNO
Lagertemperatur: Raumtemperatur
MDL: MFCD06213625
CAS-Nummer: 148870-38-6
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR